siRNA Details
| Gene Symbol: Olfr961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146504.1 | NP_666715.1 | 1 | 300 | ||
| GenBank mRNA | BC127985.1 | - | 1 | 304 | 40135556 | |
| Gene Symbol: Olfr963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011827.1 | NP_001011827.1 | 1 | 300 | ||

Olfr961
Genomic Map