siRNA Details
| Gene Symbol: Olfr148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146505.1 | NP_666716.1 | 1 | 760 | ||
| GenBank mRNA | BC125353.1 | - | 1 | 785 | 40129868 | |
| BC132239.1 | - | 1 | 785 | 40130516 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr148| Gene Symbol: Olfr148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146505.1 | NP_666716.1 | 1 | 760 | ||
| GenBank mRNA | BC125353.1 | - | 1 | 785 | 40129868 | |
| BC132239.1 | - | 1 | 785 | 40130516 | ||