siRNA Details
| Gene Symbol: Olfr945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146506.1 | NP_666717.1 | 1 | 910 | ||
| GenBank mRNA | BC128004.1 | - | 1 | 914 | 40135946 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr945| Gene Symbol: Olfr945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146506.1 | NP_666717.1 | 1 | 910 | ||
| GenBank mRNA | BC128004.1 | - | 1 | 914 | 40135946 | |