siRNA Details
| Gene Symbol: Olfr959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146508.1 | NP_666719.1 | 1 | 122 | ||
| GenBank mRNA | BC120617.1 | - | 1 | 179 | 40129540 | |
| BC120619.1 | - | 1 | 179 | 40129542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr959| Gene Symbol: Olfr959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146508.1 | NP_666719.1 | 1 | 122 | ||
| GenBank mRNA | BC120617.1 | - | 1 | 179 | 40129540 | |
| BC120619.1 | - | 1 | 179 | 40129542 | ||