siRNA Details
| Gene Symbol: Olfr523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146518.1 | NP_666729.1 | 1 | 670 | ||
| GenBank mRNA | BC119240.1 | - | 1 | 713 | 8733989 | |
| BC120529.1 | - | 1 | 713 | 40129452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr523| Gene Symbol: Olfr523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146518.1 | NP_666729.1 | 1 | 670 | ||
| GenBank mRNA | BC119240.1 | - | 1 | 713 | 8733989 | |
| BC120529.1 | - | 1 | 713 | 40129452 | ||