siRNA Details
Gene Symbol: Olfr530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146519.1 | NP_666730.1 | 1 | 574 | ||
GenBank mRNA | BC138081.1 | - | 1 | 632 | 8861100 | |
BC138082.1 | - | 1 | 632 | 8861101 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146519.1 | NP_666730.1 | 1 | 574 | ||
GenBank mRNA | BC138081.1 | - | 1 | 632 | 8861100 | |
BC138082.1 | - | 1 | 632 | 8861101 |