siRNA Details
| Gene Symbol: Olfr536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146520.2 | NP_666731.1 | 4 | 1011 | ||
| GenBank mRNA | BC051116.1 | - | 4 | 1011 | 6307473 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr536| Gene Symbol: Olfr536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146520.2 | NP_666731.1 | 4 | 1011 | ||
| GenBank mRNA | BC051116.1 | - | 4 | 1011 | 6307473 | |