siRNA Details
Gene Symbol: Olfr536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146520.2 | NP_666731.1 | 4 | 1068 | ||
GenBank mRNA | BC051116.1 | - | 4 | 1068 | 6307473 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146520.2 | NP_666731.1 | 4 | 1068 | ||
GenBank mRNA | BC051116.1 | - | 4 | 1068 | 6307473 |