siRNA Details
| Gene Symbol: Olfr850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146523.1 | NP_666734.1 | 1 | 204 | ||
| GenBank mRNA | BC113136.1 | - | 1 | 203 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr850| Gene Symbol: Olfr850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146523.1 | NP_666734.1 | 1 | 204 | ||
| GenBank mRNA | BC113136.1 | - | 1 | 203 | ||