siRNA Details
| Gene Symbol: Olfr855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146524.2 | NP_666735.2 | 1 | 483 | ||
| GenBank mRNA | BC137820.1 | - | 1 | 483 | 8860839 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr855| Gene Symbol: Olfr855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146524.2 | NP_666735.2 | 1 | 483 | ||
| GenBank mRNA | BC137820.1 | - | 1 | 483 | 8860839 | |