siRNA Details
| Gene Symbol: Olfr1186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146530.2 | NP_666741.2 | 1 | 693 | ||
| GenBank mRNA | BC132093.1 | - | 1 | 693 | 40130370 | |
| BC132095.1 | - | 1 | 693 | 40130372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1186| Gene Symbol: Olfr1186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146530.2 | NP_666741.2 | 1 | 693 | ||
| GenBank mRNA | BC132093.1 | - | 1 | 693 | 40130370 | |
| BC132095.1 | - | 1 | 693 | 40130372 | ||