siRNA Details
| Gene Symbol: Olfr1170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146532.1 | NP_666743.1 | 1 | 46 | ||
| GenBank mRNA | BC127994.1 | - | 1 | 50 | 40135743 | |
| Gene Symbol: Olfr1166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146650.2 | NP_666861.2 | 1 | 56 | ||
| GenBank mRNA | BC051250.1 | - | 1 | 56 | 6306097 | |

Olfr1170
Genomic Map