siRNA Details
| Gene Symbol: Olfr1367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146533.1 | NP_666744.1 | 1 | 592 | ||
| GenBank mRNA | BC138110.1 | - | 1 | 612 | 8861129 | |
| BC138111.1 | - | 1 | 612 | 8861130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1367| Gene Symbol: Olfr1367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146533.1 | NP_666744.1 | 1 | 592 | ||
| GenBank mRNA | BC138110.1 | - | 1 | 612 | 8861129 | |
| BC138111.1 | - | 1 | 612 | 8861130 | ||