siRNA Details
| Gene Symbol: Olfr1370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146535.1 | NP_666746.1 | 1 | 664 | ||
| GenBank mRNA | BC110468.1 | - | 1 | 664 | 40043199 | |
| BC110469.1 | - | 1 | 663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1370| Gene Symbol: Olfr1370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146535.1 | NP_666746.1 | 1 | 664 | ||
| GenBank mRNA | BC110468.1 | - | 1 | 664 | 40043199 | |
| BC110469.1 | - | 1 | 663 | |||