siRNA Details
| Gene Symbol: Olfr313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146536.2 | NP_666747.2 | 1 | 553 | ||
| GenBank mRNA | BC117002.1 | - | 1 | 553 | 40126321 | |
| BC120742.1 | - | 1 | 553 | 40129665 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr313| Gene Symbol: Olfr313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146536.2 | NP_666747.2 | 1 | 553 | ||
| GenBank mRNA | BC117002.1 | - | 1 | 553 | 40126321 | |
| BC120742.1 | - | 1 | 553 | 40129665 | ||