siRNA Details
| Gene Symbol: Olfr373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146539.2 | NP_666750.2 | 1 | 808 | ||
| GenBank mRNA | BC106979.2 | - | 1 | 806 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr373| Gene Symbol: Olfr373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146539.2 | NP_666750.2 | 1 | 808 | ||
| GenBank mRNA | BC106979.2 | - | 1 | 806 | ||