siRNA Details
| Gene Symbol: Olfr1360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146543.2 | NP_666754.2 | 1 | 361 | ||
| GenBank mRNA | BC130243.1 | - | 1 | 327 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1360| Gene Symbol: Olfr1360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146543.2 | NP_666754.2 | 1 | 361 | ||
| GenBank mRNA | BC130243.1 | - | 1 | 327 | ||