siRNA Details
| Gene Symbol: Olfr777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146544.1 | NP_666755.1 | 1 | 609 | ||
| GenBank mRNA | BC104276.1 | - | 1 | 609 | 40041253 | |
| BC104277.1 | - | 1 | 604 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr777| Gene Symbol: Olfr777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146544.1 | NP_666755.1 | 1 | 609 | ||
| GenBank mRNA | BC104276.1 | - | 1 | 609 | 40041253 | |
| BC104277.1 | - | 1 | 604 | |||