siRNA Details
| Gene Symbol: Olfr777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146544.1 | NP_666755.1 | 1 | 896 | ||
| GenBank mRNA | BC104276.1 | - | 1 | 896 | 40041253 | |
| BC104277.1 | - | 1 | 891 | |||
| Gene Symbol: Olfr800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146548.1 | NP_666759.1 | 1 | 896 | ||
| GenBank mRNA | BC134373.1 | - | 1 | 900 | 40134856 | |

Olfr777
Genomic Map