siRNA Details
| Gene Symbol: Olfr775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146545.2 | NP_666756.2 | 1 | 875 | ||
| Gene Symbol: Olfr774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207620.1 | NP_997503.1 | 1 | 875 | ||
| Gene Symbol: Olfr776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207559.1 | NP_997442.1 | 1 | 875 | ||

Olfr775
Genomic Map