siRNA Details
| Gene Symbol: Olfr868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146559.2 | NP_666770.1 | 2 | 317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr868| Gene Symbol: Olfr868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146559.2 | NP_666770.1 | 2 | 317 | ||