siRNA Details
Gene Symbol: Olfr837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146565.2 | NP_666776.1 | 1 | 387 | ||
GenBank mRNA | BC140981.1 | - | 1 | 387 | 9055593 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146565.2 | NP_666776.1 | 1 | 387 | ||
GenBank mRNA | BC140981.1 | - | 1 | 387 | 9055593 |