siRNA Details
Gene Symbol: Olfr843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146567.2 | NP_666778.2 | 1 | 832 | ||
GenBank mRNA | BC138386.1 | - | 1 | 832 | 8861406 | |
BC138387.1 | - | 1 | 832 | 8861407 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146567.2 | NP_666778.2 | 1 | 832 | ||
GenBank mRNA | BC138386.1 | - | 1 | 832 | 8861406 | |
BC138387.1 | - | 1 | 832 | 8861407 |