siRNA Details
Gene Symbol: Olfr1012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146568.2 | NP_666779.1 | 1 | 79 | ||
GenBank mRNA | BC111581.1 | - | 1 | 77 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146568.2 | NP_666779.1 | 1 | 79 | ||
GenBank mRNA | BC111581.1 | - | 1 | 77 |