siRNA Details
| Gene Symbol: Olfr1014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146569.2 | NP_666780.2 | 1 | 511 | ||
| Gene Symbol: Olfr1013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146762.2 | NP_666973.2 | 1 | 511 | ||
| GenBank mRNA | DR065625.1 | - | 555 | |||

Olfr1014
Genomic Map