siRNA Details
Gene Symbol: Olfr1009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146572.2 | NP_666783.1 | 1 | 133 | ||
GenBank mRNA | BC111578.1 | - | 1 | 132 | ||
BC148238.1 | - | 1 | 133 | 40042413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146572.2 | NP_666783.1 | 1 | 133 | ||
GenBank mRNA | BC111578.1 | - | 1 | 132 | ||
BC148238.1 | - | 1 | 133 | 40042413 |