siRNA Details
| Gene Symbol: Olfr1033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146578.2 | NP_666789.1 | 3 | 501 | ||
| GenBank mRNA | AK035445.1 | - | 2 | 383 | ||
| AK041245.1 | - | 3 | 501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1033| Gene Symbol: Olfr1033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146578.2 | NP_666789.1 | 3 | 501 | ||
| GenBank mRNA | AK035445.1 | - | 2 | 383 | ||
| AK041245.1 | - | 3 | 501 | |||