siRNA Details
| Gene Symbol: Olfr1032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146579.2 | NP_666790.2 | 1 | 223 | ||
| GenBank mRNA | BC113200.1 | - | 1 | 222 | ||
| Gene Symbol: Olfr1033 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146578.2 | NP_666789.1 | 3 | 475 | ||
| GenBank mRNA | AK035445.1 | - | 2 | 357 | ||
| AK041245.1 | - | 3 | 475 | |||

Olfr1032
Genomic Map