siRNA Details
Gene Symbol: Olfr1022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146589.2 | NP_666800.2 | 1 | 618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146589.2 | NP_666800.2 | 1 | 618 |