siRNA Details
| Gene Symbol: Olfr1111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146593.2 | NP_666804.2 | 1 | 49 | ||
| GenBank mRNA | BC104068.2 | - | 1 | 47 | ||
| BC104069.1 | - | 1 | 49 | 40041592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1111| Gene Symbol: Olfr1111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146593.2 | NP_666804.2 | 1 | 49 | ||
| GenBank mRNA | BC104068.2 | - | 1 | 47 | ||
| BC104069.1 | - | 1 | 49 | 40041592 | ||