siRNA Details
| Gene Symbol: Olfr1100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146594.1 | NP_666805.1 | 1 | 405 | ||
| GenBank mRNA | BC130249.1 | - | 1 | 409 | 40142420 | |
| BC139802.1 | - | 1 | 409 | 40142396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1100| Gene Symbol: Olfr1100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146594.1 | NP_666805.1 | 1 | 405 | ||
| GenBank mRNA | BC130249.1 | - | 1 | 409 | 40142420 | |
| BC139802.1 | - | 1 | 409 | 40142396 | ||