siRNA Details
| Gene Symbol: Olfr698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146602.2 | NP_666813.2 | 1 | 407 | ||
| GenBank mRNA | BC113955.1 | - | 1 | 406 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr698| Gene Symbol: Olfr698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146602.2 | NP_666813.2 | 1 | 407 | ||
| GenBank mRNA | BC113955.1 | - | 1 | 406 | ||