siRNA Details
| Gene Symbol: Olfr270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146607.1 | NP_666818.1 | 1 | 670 | ||
| GenBank mRNA | BC127971.1 | - | 1 | 674 | 40135114 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr270| Gene Symbol: Olfr270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146607.1 | NP_666818.1 | 1 | 670 | ||
| GenBank mRNA | BC127971.1 | - | 1 | 674 | 40135114 | |