siRNA Details
Gene Symbol: Olfr970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146611.1 | NP_666822.1 | 1 | 489 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146611.1 | NP_666822.1 | 1 | 489 |