siRNA Details
| Gene Symbol: Olfr986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146615.2 | NP_666826.1 | 1 | 866 | ||
| GenBank mRNA | BC116768.1 | - | 1 | 866 | 40126087 | |
| BC116770.1 | - | 1 | 866 | 40126089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr986| Gene Symbol: Olfr986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146615.2 | NP_666826.1 | 1 | 866 | ||
| GenBank mRNA | BC116768.1 | - | 1 | 866 | 40126087 | |
| BC116770.1 | - | 1 | 866 | 40126089 | ||