siRNA Details
| Gene Symbol: Olfr308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146621.1 | NP_666832.1 | 1 | 396 | ||
| GenBank mRNA | BC119213.1 | - | 1 | 496 | 8733962 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr308| Gene Symbol: Olfr308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146621.1 | NP_666832.1 | 1 | 396 | ||
| GenBank mRNA | BC119213.1 | - | 1 | 496 | 8733962 | |