siRNA Details
| Gene Symbol: Olfr356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146624.1 | NP_666835.1 | 1 | 68 | ||
| GenBank mRNA | BC129799.1 | - | 1 | 68 | 40042987 | |
| BC148201.1 | - | 1 | 68 | 40042961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr356| Gene Symbol: Olfr356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146624.1 | NP_666835.1 | 1 | 68 | ||
| GenBank mRNA | BC129799.1 | - | 1 | 68 | 40042987 | |
| BC148201.1 | - | 1 | 68 | 40042961 | ||