siRNA Details
Gene Symbol: Olfr356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146624.1 | NP_666835.1 | 1 | 94 | ||
GenBank mRNA | BC129799.1 | - | 1 | 94 | 40042987 | |
BC148201.1 | - | 1 | 94 | 40042961 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146624.1 | NP_666835.1 | 1 | 94 | ||
GenBank mRNA | BC129799.1 | - | 1 | 94 | 40042987 | |
BC148201.1 | - | 1 | 94 | 40042961 |