siRNA Details
| Gene Symbol: Olfr120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146631.1 | NP_666842.1 | 1 | 274 | ||
| Gene Symbol: Olfr118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213721.2 | NP_998886.2 | 1 | 247 | ||
| GenBank mRNA | BC128012.1 | - | 1 | 251 | 40136025 | |

Olfr120
Genomic Map