siRNA Details
| Gene Symbol: Olfr1504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146634.1 | NP_666845.1 | 1 | 212 | ||
| GenBank mRNA | BC137835.1 | - | 1 | 309 | 8860854 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1504| Gene Symbol: Olfr1504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146634.1 | NP_666845.1 | 1 | 212 | ||
| GenBank mRNA | BC137835.1 | - | 1 | 309 | 8860854 | |