siRNA Details
| Gene Symbol: Olfr1163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146644.2 | NP_666855.1 | 1 | 493 | ||
| GenBank mRNA | BC120531.1 | - | 1 | 493 | 40129454 | |
| BC120557.1 | - | 1 | 493 | 40129480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1163| Gene Symbol: Olfr1163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146644.2 | NP_666855.1 | 1 | 493 | ||
| GenBank mRNA | BC120531.1 | - | 1 | 493 | 40129454 | |
| BC120557.1 | - | 1 | 493 | 40129480 | ||