siRNA Details
| Gene Symbol: Olfr1166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146650.2 | NP_666861.2 | 1 | 617 | ||
| GenBank mRNA | BC051250.1 | - | 1 | 617 | 6306097 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1166| Gene Symbol: Olfr1166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146650.2 | NP_666861.2 | 1 | 617 | ||
| GenBank mRNA | BC051250.1 | - | 1 | 617 | 6306097 | |