siRNA Details
Gene Symbol: Olfr444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146656.1 | NP_666867.1 | 1 | 292 | ||
GenBank mRNA | BC145664.1 | - | 1 | 333 | 40130967 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146656.1 | NP_666867.1 | 1 | 292 | ||
GenBank mRNA | BC145664.1 | - | 1 | 333 | 40130967 |