siRNA Details
| Gene Symbol: Olfr734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146664.1 | NP_666875.1 | 1 | 127 | ||
| GenBank mRNA | BC141029.1 | - | 1 | 162 | 9055641 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr734| Gene Symbol: Olfr734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146664.1 | NP_666875.1 | 1 | 127 | ||
| GenBank mRNA | BC141029.1 | - | 1 | 162 | 9055641 | |