siRNA Details
Gene Symbol: Olfr738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146420.2 | NP_666532.2 | 1 | 591 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
