siRNA Details
| Gene Symbol: Olfr739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146668.2 | NP_666879.2 | 1 | 823 | ||
| Gene Symbol: Olfr741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207133.2 | NP_997016.2 | 3 | 1037 | ||
| Gene Symbol: Olfr742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146430.2 | NP_666641.2 | 1 | 914 | ||
| GenBank mRNA | BC132583.1 | - | 1 | 914 | 40130860 | |
More Information
Set Membership: |
|

Olfr739
Genomic Map