siRNA Details
| Gene Symbol: Olfr816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146672.1 | NP_666883.1 | 1 | 733 | ||
| GenBank mRNA | BC100682.1 | - | 1 | 731 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr816| Gene Symbol: Olfr816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146672.1 | NP_666883.1 | 1 | 733 | ||
| GenBank mRNA | BC100682.1 | - | 1 | 731 | ||