siRNA Details
| Gene Symbol: Olfr823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146673.2 | NP_666884.2 | 1 | 707 | ||
| Gene Symbol: Olfr824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146674.1 | NP_666885.1 | 1 | 707 | ||
| GenBank mRNA | BC104094.1 | - | 1 | 699 | ||
| BC104095.2 | - | 1 | 699 | |||
| BC106800.2 | - | 1 | 699 | |||

Olfr823
Genomic Map