siRNA Details
| Gene Symbol: Olfr824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146674.1 | NP_666885.1 | 1 | 148 | ||
| GenBank mRNA | BC104094.1 | - | 1 | 140 | ||
| BC104095.2 | - | 1 | 140 | |||
| BC106800.2 | - | 1 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr824| Gene Symbol: Olfr824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146674.1 | NP_666885.1 | 1 | 148 | ||
| GenBank mRNA | BC104094.1 | - | 1 | 140 | ||
| BC104095.2 | - | 1 | 140 | |||
| BC106800.2 | - | 1 | 140 | |||