siRNA Details
| Gene Symbol: Olfr825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146677.1 | NP_666888.1 | 1 | 913 | ||
| GenBank mRNA | BC119054.1 | - | 1 | 1011 | 8733803 | |
| BC119056.1 | - | 1 | 1011 | 8733805 | ||
| Gene Symbol: Olfr826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146676.1 | NP_666887.1 | 1 | 913 | ||

Olfr825
Genomic Map